Benzoyl-DL-leucine ≥99.0% (by HPLC, titration analysis)
Proveedor: TCI
B0206-10G
B0206-25G
TCIAB0206-10GEA
26
EUR
TCIAB0206-10G
TCIAB0206-25G
Benzoyl-DL-leucine ≥99.0% (by HPLC, titration analysis)
Benzoyl-DL-leucine
Formula:
C₁₃H₁₇NO₃ |
Número MDL:
MFCD00038295 Núm. CAS: 17966-67-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Neutralization titration) | min. 99.0 % |
Solubility in Methanol | almost transparency |
Melting point | 139.0 to 143.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...