Benzoyl leuco methylene blue ≥96.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
(3,7-Bis(dimethylamino)-10H-phenothiazin-10-yl)(phenyl)methanone
B0207-1G
B0207-25G
TCIAB0207-1GEA
16
EUR
TCIAB0207-1G
TCIAB0207-25G
Benzoyl leuco methylene blue ≥96.0% (by HPLC, titration analysis)
Benzoyl leuco methylene blue
Formula:
C₂₃H₂₃N₃OS MW: 389,52 g/mol |
Número MDL:
MFCD00059153 Núm. CAS: 1249-97-4 EINECS: 215-005-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Solubility in hot Toluene | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...