Ácido naftaleno-2-sulfónico ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Peligro
|
B0258-25G
B0258-500G
TCIAB0258-25GEA
16
EUR
TCIAB0258-25G
TCIAB0258-500G
Ácido naftaleno-2-sulfónico ≥98.0% (by HPLC, titration analysis)
Ácido naftaleno-2-sulfónico
Formula:
C₁₀H₈O₃S |
Número MDL:
MFCD00149271 Núm. CAS: 120-18-3 EINECS: 204-375-3 UN: 2585 ADR: 8,III Índice Merck: 14,06377 |
Resultados test especificaciones
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd. on anh.substance) |
Water | max. 10.0 % |
Sulfuric acid | max. 0.2 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...