L-serine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI

B0267-25G B0267-5G
TCIAB0267-25GEA 76 EUR
TCIAB0267-25G TCIAB0267-5G
L-serine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)
L-serine methyl ester hydrochloride
Formula: C₄H₉NO₃
Número MDL: MFCD00063680
Núm. CAS: 5680-80-8
EINECS: 227-140-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=844e4213c6a45d7e9d360c50be89a36b92120548&vid=844e4213c6a45d7e9d360c50be89a36b92120548&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +2.0 to +5.0 deg (C=5,MeOH)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR