2-Naphthyl phenyl ketone ≥98.0% (por GC)

Proveedor: TCI

B0301-25G B0301-5G
TCIAB0301-25GEA 317 EUR
TCIAB0301-25G TCIAB0301-5G
2-Naphthyl phenyl ketone ≥98.0% (por GC)
2-Naphthyl phenyl ketone
Formula: C₁₇H₁₂O
Pto. Ebullición: 225°C
Número MDL: MFCD00004106
Núm. CAS: 644-13-3
EINECS: 211-410-6

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 84.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR