Benzoyl-DL-valine ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Benzoyl-DL-Val-OH, 2-Benzamido-3-methylbutanoic acid

B0401-1G B0401-25G
TCIAB0401-1GEA 16 EUR
TCIAB0401-1G TCIAB0401-25G
Benzoyl-DL-valine ≥98.0% (by titrimetric analysis)
Benzoyl-DL-valine
Formula: C₁₂H₁₅NO₃
MW: 221,26 g/mol
Número MDL: MFCD00038283
Núm. CAS: 2901-80-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 131.0 to 133.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR