Diethyl benzylidenemalonate ≥96.0% (por GC)
Proveedor: TCI
Sinónimos:
Diethyl Benzalmalonate, Benzalmalonic Acid Diethyl Ester, Benzylidenemalonic Acid Diethyl Ester
B0420-25G
B0420-5G
TCIAB0420-25GEA
34
EUR
TCIAB0420-25G
TCIAB0420-5G
Diethyl benzylidenemalonate ≥96.0% (por GC)
Diethyl benzylidenemalonate
Formula:
C₁₄H₁₆O₄ MW: 248,28 g/mol |
Número MDL:
MFCD00009149 Núm. CAS: 5292-53-5 EINECS: 226-137-8 |
Resultados test especificaciones
Appearance | Colorless to Yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...