3,6-Bis(hydroxymethyl)durene ≥99.0% (por GC)
Proveedor: TCI
Sinónimos:
2,3,5,6-Tetramethyl-p-xylene-alpha,alpha'-diol, 1,4-Bis(hydroxymethyl)-2,3,5,6-tetramethylbenzene
B0493-1G
B0493-5G
TCIAB0493-1GEA
64
EUR
TCIAB0493-1G
TCIAB0493-5G
3,6-Bis(hydroxymethyl)durene ≥99.0% (por GC)
3,6-Bis(hydroxymethyl)durene
Formula:
C₁₂H₁₈O₂ MW: 194,27 g/mol |
Número MDL:
MFCD00004623 Núm. CAS: 7522-62-5 EINECS: 231-382-9 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 254.0 to 258.0 deg-C |
Solubility in hot Pyridine | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...