N-tert-Butylacrylamide ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
B0703-25G
B0703-100G
TCIAB0703-25GEA
20
EUR
TCIAB0703-25G
TCIAB0703-100G
N-tert-Butylacrylamide ≥98.0% (by GC, titration analysis)
N-tert-Butylacrylamide
Formula:
C₇H₁₃NO |
Número MDL:
MFCD00026271 Núm. CAS: 107-58-4 EINECS: 203-505-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Bromination Method) | min. 98.0 % |
Melting point | 127.0 to 131.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...