Benzopurpurine 4B

Proveedor: TCI

Sinónimos: Direct Red 2, CI 23500, Direct Red 3, Benzopurpurin B

B0781-25G B0781-5G
TCIAB0781-25GEA 1121 EUR
TCIAB0781-25G TCIAB0781-5G
Benzopurpurine 4B
Benzopurpurine 4B
Formula: C₃₄H₂₈N₆O₆S₂
MW: 724,72 g/mol
Número MDL: MFCD00012410
Núm. CAS: 992-59-6
EINECS: 213-594-3
Índice Merck: 14,01101
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2414dcea6850c9f420bd66340f0ece7e3b12668f&vid=df41b3772dc1a6e4f9312e9d174df5f35eecf6cd&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Orange to Brown powder to crystal
Lambda max. 480.0 to 495.0 nm (pH 4.4 buffer sol.)
Absorbance min. 0.850 (20mg/L, pH 4.4 buffer sol., 480.0 to 495.0 nm) (calcd.on dried substance)
Drying loss max. 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR