S-Benzyl-L-cysteine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: H-Cys(Bzl)-OH

B0865-1G B0865-25G B0865-5G
TCIAB0865-1GEA 16 EUR
TCIAB0865-1G TCIAB0865-25G TCIAB0865-5G
S-Benzyl-L-cysteine ≥98.0% (by HPLC, titration analysis)
S-Benzyl-L-cysteine
Formula: C₁₀H₁₃NO₂S
MW: 211,28 g/mol
Número MDL: MFCD00002613
Núm. CAS: 3054-01-1
EINECS: 221-273-4

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +27.5 to +30.5 deg (C=1, 1mol/L NaOH sol.)
Solubility in 1mol/L NaOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR