Terephthalbis(p-phenetidine) ≥98.0% (by GC, total nitrogen)

Proveedor: TCI

Sinónimos: Di-p-phenetidylterephthalaldehyde, Bisphenetidylterephthalaldehyde, Bis(p-ethoxyanilino)terephthalaldehyde

B0871-25G B0871-5G
TCIAB0871-25GEA 216 EUR
TCIAB0871-25G TCIAB0871-5G
Terephthalbis(p-phenetidine) ≥98.0% (by GC, total nitrogen)
Terephthalbis(p-phenetidine)
Formula: C₂₄H₂₄N₂O₂
MW: 372,47 g/mol
Número MDL: MFCD00026796
Núm. CAS: 17696-60-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR