4-Benzoyl-3-methyl-1-phenyl-5-pyrazolone ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 4-Benzoyl-5-methyl-2-phenyl-2,4-dihydro-3H-pyrazol-3-one

B0976-25G B0976-5G
TCIAB0976-25GEA 339 EUR
TCIAB0976-25G TCIAB0976-5G
4-Benzoyl-3-methyl-1-phenyl-5-pyrazolone ≥98.0% (by HPLC, titration analysis)
4-Benzoyl-3-methyl-1-phenyl-5-pyrazolone
Formula: C₁₇H₁₄N₂O₂
MW: 278,31 g/mol
Número MDL: MFCD00003136
Núm. CAS: 4551-69-3
EINECS: 224-918-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 116.0 to 120.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR