2,2'-Bicinchoninic acid dipotassium salt hydrate ≥98.0% (by HPLC) for determination of Cu
Proveedor: TCI
|
Atención
|
Sinónimos:
4,4'-Dicarboxy-2,2'-biquinoline Dipotassium Salt, Potassium [2,2'-biquinoline]-4,4'-dicarboxylate, 2,2'-Biquinoline-4,4'-dicarboxylic Acid Dipotassium Salt
B1077-1G
B1077-5G
TCIAB1077-1GEA
31
EUR
TCIAB1077-1G
TCIAB1077-5G
2,2'-Bicinchoninic acid dipotassium salt hydrate ≥98.0% (by HPLC) for determination of Cu
2,2'-Bicinchoninic acid dipotassium salt hydrate
Formula:
C₂₀H₁₂N₂O₄ |
Número MDL:
MFCD00014664 Núm. CAS: 63451-34-3 EINECS: 264-196-1 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Lambda max. | 261.0 to 264.0 nm (EtOH) |
Absorbance (E1%1cm) | min. 750 (5mg/L, ethanol, 261.0 to 264.0nm) |
Drying loss | 5.0 to 15.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...