2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine ≥98.0% (by titrimetric analysis) BOC agent for peptides synthesis
Proveedor: TCI
Sinónimos:
2-Boc-thio-4,6-dimethylpyrimidine, O-tert-Butyl S-(4,6-dimethylpyrimidin-2-yl) carbonothioate
B1089-25G
B1089-5G
TCIAB1089-25GEA
100
EUR
TCIAB1089-25G
TCIAB1089-5G
2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine ≥98.0% (by titrimetric analysis) BOC agent for peptides synthesis
2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine
Formula:
C₁₁H₁₆N₂O₂S MW: 240,32 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00006080 Núm. CAS: 41840-28-2 EINECS: 255-562-1 |
Resultados test especificaciones
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 50.0 to 54.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...