Bis(4-nitrophenyl)phosphate ≥98.0% (by titrimetric analysis) for phosphodiesterase substrate

Proveedor: TCI
Atención

B1098-1G B1098-10G
TCIAB1098-1GEA 127 EUR
TCIAB1098-1G TCIAB1098-10G
Bis(4-nitrophenyl)phosphate ≥98.0% (by titrimetric analysis) for phosphodiesterase substrate
Bis(4-nitrophenyl)phosphate
Formula: C₁₂H₉N₂O₈P
Número MDL: MFCD00007318
Núm. CAS: 645-15-8
EINECS: 211-434-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 175.0 to 179.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR