(-)-cis-2-Benzylaminocyclohexanemethanol ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: (-)-N-Benzyl-cis-2-hydroxymethylcyclohexylamine

B1118-1G B1118-5G
TCIAB1118-1GEA 73 EUR
TCIAB1118-1G TCIAB1118-5G
(-)-cis-2-Benzylaminocyclohexanemethanol ≥98.0% (by titrimetric analysis)
(-)-cis-2-Benzylaminocyclohexanemethanol
Formula: C₁₄H₂₁NO
MW: 219,33 g/mol
Número MDL: MFCD00151396
Núm. CAS: 71581-93-6

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 66.0 to 69.0 deg-C
Specific rotation [a]20/D -22.0 to -26.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR