5-Mercapto-3-phenyl-1,3,4-thiadiazole-2(3H)thione potassium salt ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: Bismuthiol II, 5-Mercapto-3-phenyl-1,3,4-thiadiazole-2-thione Potassium Salt

B1124-25G B1124-5G
TCIAB1124-25GEA 109 EUR
TCIAB1124-25G TCIAB1124-5G
5-Mercapto-3-phenyl-1,3,4-thiadiazole-2(3H)thione potassium salt ≥98.0% (by HPLC, titration analysis)
5-Mercapto-3-phenyl-1,3,4-thiadiazole-2(3H)thione potassium salt
Formula: C₈H₅KN₂S₃
Temperatura de almacenaje: Nevera
Número MDL: MFCD00014092
Núm. CAS: 6336-51-2
EINECS: 228-718-1

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on dried substance)
Solubility in Water almost transparency
Drying loss max. 8.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR