N-Benzoyl-L-tyrosine ethyl ester ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

B1140-1G B1140-5G
TCIAB1140-1GEA 21 EUR
TCIAB1140-1G TCIAB1140-5G
N-Benzoyl-L-tyrosine ethyl ester ≥98.0% (by HPLC, total nitrogen)
N-Benzoyl-L-tyrosine ethyl ester
Formula: C₁₈H₁₉NO₄
Número MDL: MFCD00002388
Núm. CAS: 3483-82-7
EINECS: 222-469-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 120.0 to 124.0 deg-C
Specific rotation [a]20/D -25.0 to -30.0 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR