Boc-L-leucine monohydrate ≥99.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Boc-Leu-OH·H₂O, Boc-Leu-OH·H2O, N-(tert-Butoxycarbonyl)-L-leucine monohydrate
B1187-25G
B1187-5G
TCIAB1187-25GEA
45
EUR
TCIAB1187-25G
TCIAB1187-5G
Boc-L-leucine monohydrate ≥99.0% (by titrimetric analysis)
Boc-L-leucine monohydrate
Formula:
C₁₁H₂₁NO₄·H₂O MW: 249,307 g/mol |
Número MDL:
MFCD00065582 Núm. CAS: 200936-87-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 85.0 to 88.0 deg-C |
Specific rotation [a]20/D | -24.0 to -26.0 deg (C=2, AcOH) |
Solubility in Acetic acid | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...