Benzyldimethylstearylammonium chloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

B1297-25G B1297-100G
TCIAB1297-25GEA 29 EUR
TCIAB1297-25G TCIAB1297-100G
Benzyldimethylstearylammonium chloride ≥98.0% (by titrimetric analysis)
Benzyldimethylstearylammonium chloride
Formula: C₂₇H₅₀ClN
Número MDL: MFCD00011776
Núm. CAS: 122-19-0
EINECS: 204-527-9

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 90.0 area%
Purity (TPBNa Method) min. 98.0 % (calcd.on anh.substance)
Water max. 12.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR