9,10-Bis(chloromethyl)anthracene ≥97.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

B1327-25G B1327-5G
TCIAB1327-25GEA 382 EUR
TCIAB1327-25G TCIAB1327-5G
9,10-Bis(chloromethyl)anthracene ≥97.0% (by GC, titration analysis)
9,10-Bis(chloromethyl)anthracene
Formula: C₁₆H₁₂Cl₂
MW: 275.17 g/mol
Número MDL: MFCD00045388
Núm. CAS: 10387-13-0
UN: 3261
ADR: 8 II

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %
Solubility in hot Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR