Ethyl-3,3-bis(methylthio)-2-cyanoacrylate ≥98.0% (by total nitrogen basis)

Proveedor: TCI
Atención

Sinónimos: 3,3-Bis(methylthio)-2-cyanoacrylic Acid Ethyl Ester, Ethyl 2-cyano-3,3-bis(methylthio)acrylate

B1411.0500 B1411-25G B1411-500G B1411-100G B1411-5G
TCIAB1411.0500EA 0 EUR
TCIAB1411.0500 TCIAB1411-25G TCIAB1411-500G TCIAB1411-100G TCIAB1411-5G
Ethyl-3,3-bis(methylthio)-2-cyanoacrylate ≥98.0% (by total nitrogen basis)
Ethyl-3,3-bis(methylthio)-2-cyanoacrylate
Formula: C₈H₁₁NO₂S₂
MW: 217,30 g/mol
Número MDL: MFCD00143027
Núm. CAS: 17823-58-4

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 56.0 to 59.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR