2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4,4'-(Hexafluoroisopropylidene)bis(2-aminophenol), 4,4'-(Perfluoropropane-2,2-diyl)bis(2-aminophenol)
B1415-25G
B1415-5G
TCIAB1415-25GEA
74
EUR
TCIAB1415-25G
TCIAB1415-5G
2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane ≥98.0% (by titrimetric analysis)
2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane
Formula:
C₁₅H₁₂F₆N₂O₂ MW: 366,26 g/mol |
Número MDL:
MFCD00039147 Núm. CAS: 83558-87-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...