Butoxyacetic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

B1467-1G B1467-5G
TCIAB1467-1GEA 43 EUR
TCIAB1467-1G TCIAB1467-5G
Butoxyacetic acid ≥98.0% (by GC, titration analysis)
Butoxyacetic acid
Formula: C₆H₁₂O₃
Pto. Ebullición: 234°C
Número MDL: MFCD00067044
Núm. CAS: 2516-93-0
UN: 3265
ADR: 8,III

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR