ɑ,ɑ'-Bis(4-methylbenzyloxy)-p-xylene ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
p-Xylylene Glycol Bis(4-methylbenzyl) Ether, α,α'-Bis(4-methylbenzyloxy)-p-xylene
B1482-1G
B1482-5G
TCIAB1482-1GEA
145
EUR
TCIAB1482-1G
TCIAB1482-5G
ɑ,ɑ'-Bis(4-methylbenzyloxy)-p-xylene ≥98.0% (por GC)
alpha,alpha'-Bis(4-methylbenzyloxy)-p-xylene
Formula:
C₂₄H₂₆O₂ MW: 346,47 g/mol |
Número MDL:
MFCD00143577 Núm. CAS: 136861-46-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 99.0 to 102.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...