2,3-Bis(2,4,5-trimethyl-3-thienyl)maleic anhydride ≥97.0% (by titrimetric analysis)

Proveedor: TCI

B1534-1G B1534-100MG
TCIAB1534-1GEA 720 EUR
TCIAB1534-1G TCIAB1534-100MG
2,3-Bis(2,4,5-trimethyl-3-thienyl)maleic anhydride ≥97.0% (by titrimetric analysis)
2,3-Bis(2,4,5-trimethyl-3-thienyl)maleic anhydride
Formula: C₁₈H₁₈O₃S₂
MW: 346,46 g/mol
Número MDL: MFCD00142789
Núm. CAS: 112440-47-8

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Green powder to crystal
Purity (Methanolysis Method) min. 97.0 %
Melting point 166.0 to 169.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR