9,9-Bis(4-aminophenyl)fluorene ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 4,4'-(9-Fluorenylidene)dianiline, 4,4'-(9H-Fluorene-9,9-diyl)dianiline

B1549-25G B1549-100G
TCIAB1549-25GEA 66 EUR
TCIAB1549-25G TCIAB1549-100G
9,9-Bis(4-aminophenyl)fluorene ≥98.0% (by titrimetric analysis)
9,9-Bis(4-aminophenyl)fluorene
Formula: C₂₅H₂₀N₂
MW: 348,45 g/mol
Número MDL: MFCD00039156
Núm. CAS: 15499-84-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in N,N-DMF very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR