2,2-Bis(4-hydroxy-3-methylphenyl)propane ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
4,4'-(Propane-2,2-diyl)bis(2-methylphenol), 4,4'-Isopropylidenedi-o-cresol
B1567-25G
B1567-500G
B1567-100G
TCIAB1567-25GEA
35
EUR
TCIAB1567-25G
TCIAB1567-500G
TCIAB1567-100G
2,2-Bis(4-hydroxy-3-methylphenyl)propane ≥98.0% (por GC)
2,2-Bis(4-hydroxy-3-methylphenyl)propane
Formula:
C₁₇H₂₀O₂ MW: 256,35 g/mol |
Número MDL:
MFCD00002232 Núm. CAS: 79-97-0 EINECS: 201-240-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 139.0 to 142.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...