Bis[4-(2-hydroxyethoxy)phenyl]sulphone ≥97.0% (por GC)

Proveedor: TCI

Sinónimos: 4-(2-Hydroxyethoxy)phenyl Sulphone

B1590-25G B1590-500G
TCIAB1590-25GEA 16 EUR
TCIAB1590-25G TCIAB1590-500G
Bis[4-(2-hydroxyethoxy)phenyl]sulphone ≥97.0% (por GC)
Bis[4-(2-hydroxyethoxy)phenyl]sulphone
Formula: C₁₆H₁₈O₆S
Número MDL: MFCD00130282
Núm. CAS: 27205-03-4
EINECS: 248-320-1

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 180.0 to 185.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR