N-Boc-L-serine ≥97.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: (S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxypropanoic acid, Boc-Ser-OH, N-(tert-Butoxycarbonyl)-L-serine, Boc-L-serine, N-(tert-Butoxycarbonyl)-L-serine hydrate, BOC-L-Serine hydrate

B1637-25G B1637-5G
TCIAB1637-25GEA 83 EUR
TCIAB1637-25G TCIAB1637-5G
N-Boc-L-serine ≥97.0% (by titrimetric analysis)
N-Boc-L-serine
Formula: C₈H₁₅NO₅
MW: 205,21 g/mol
Número MDL: MFCD00037243
Núm. CAS: 3262-72-4
EINECS: 221-867-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D -3.0 to -5.0 deg (C=1, AcOH)
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR