2,2-Bis(3-aminophenyl)hexafluoropropane ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3,3'-(Hexafluoroisopropylidene)dianiline
B1704-25G
B1704-5G
TCIAB1704-25GEA
1304
EUR
TCIAB1704-25G
TCIAB1704-5G
2,2-Bis(3-aminophenyl)hexafluoropropane ≥98.0% (by GC, titration analysis)
2,2-Bis(3-aminophenyl)hexafluoropropane
Formula:
C₁₅H₁₂F₆N₂ MW: 334,27 g/mol |
Número MDL:
MFCD00191619 Núm. CAS: 47250-53-3 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 74.0 to 79.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...