O-tert-Butyl-L-serine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
H-Ser(tBu)-OMe·HCl, H-Ser(tBu)-OMe.HCl
B1736-5G
B1736-1G
TCIAB1736-5GEA
1038
EUR
TCIAB1736-5G
TCIAB1736-1G
O-tert-Butyl-L-serine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)
O-tert-Butyl-L-serine methyl ester hydrochloride
Formula:
C₈H₁₇NO₃ MW: 211,69 g/mol |
Número MDL:
MFCD00077108 Núm. CAS: 17114-97-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Specific rotation [a]20/D | +6.8 to +8.2 deg (C=1, DMF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...