meso-Butane-1,2,3,4-tetracarboxylic dianhydride ≥96.0% (by titrimetric analysis)

Proveedor: TCI
Atención

B1770-25G B1770-5G
TCIAB1770-25GEA 207 EUR
TCIAB1770-25G TCIAB1770-5G
meso-Butane-1,2,3,4-tetracarboxylic dianhydride ≥96.0% (by titrimetric analysis)
meso-Butane-1,2,3,4-tetracarboxylic dianhydride
Formula: C₈H₆O₆
Número MDL: MFCD00671500
Núm. CAS: 17309-39-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Morpholine Method) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR