β-methasone ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
9ɑ-Fluoro-11β,17ɑ,21-trihydroxy-16β-methyl-1,4-pregnadiene-3,20-dione
B1837-1G
B1837-5G
TCIAB1837-1GEA
41
EUR
TCIAB1837-1G
TCIAB1837-5G
β-methasone ≥98.0% (by HPLC)
Betamethasone
Formula:
C₂₂H₂₉FO₅ MW: 392,47 g/mol |
Número MDL:
MFCD00062969 Núm. CAS: 378-44-9 EINECS: 206-825-4 Índice Merck: 14,01180 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | +115 to +121 deg (C=1, Dioxane) |
Solubility in hot Dioxane | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...