Nε-(tert-Butoxycarbonyl)-Nɑ-carbobenzoxy-L-lysine ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
Nepsilon-Boc-Nalpha-Cbz-L-lysine, Z-Lys(Boc)-OH
B1862-25G
B1862-5G
TCIAB1862-25GEA
346
EUR
TCIAB1862-25G
TCIAB1862-5G
Nε-(tert-Butoxycarbonyl)-Nɑ-carbobenzoxy-L-lysine ≥98.0% (by HPLC, titration analysis)
Nepsilon-(tert-Butoxycarbonyl)-Nalpha-carbobenzoxy-L-lysine
Formula:
C₁₉H₂₈N₂O₆ MW: 380,44 g/mol |
Número MDL:
MFCD00062271 Núm. CAS: 2389-60-8 EINECS: 219-223-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 60.0 to 64.0 deg-C |
Specific rotation [a]20/D | -4.0 to -7.0 deg (C=1, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...