4-Biphenyl benzoate ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: Benzoic Acid 4-Biphenyl Ester, (4-Phenylphenyl) benzoate

B1866-25G B1866-5G
TCIAB1866-25GEA 400 EUR
TCIAB1866-25G TCIAB1866-5G
4-Biphenyl benzoate ≥98.0% (por GC)
4-Biphenyl benzoate
Formula: C₁₉H₁₄O₂
MW: 274,32 g/mol
Número MDL: MFCD00223541
Núm. CAS: 2170-13-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 150.0 to 153.0 deg-C
Solubility in hot Toluene very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR