Benzenesulphonic anhydride ≥96.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

B1931-25G B1931-5G
TCIAB1931-25GEA 395 EUR
TCIAB1931-25G TCIAB1931-5G
Benzenesulphonic anhydride ≥96.0% (by titrimetric analysis)
Benzenesulphonic anhydride
Formula: C₁₂H₁₀O₅S₂
MW: 298,33 g/mol
Número MDL: MFCD00054601
Núm. CAS: 512-35-6
UN: 3261
ADR: 8,III
Índice Merck: 14,01071

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (Anilin Method) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR