3-(2-Benzothiazolylthio)propionic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: 3-(Benzo[d]thiazol-2-ylthio)propanoic acid

B1942-25G B1942-500G
TCIAB1942-25GEA 17 EUR
TCIAB1942-25G TCIAB1942-500G
3-(2-Benzothiazolylthio)propionic acid ≥98.0% (by titrimetric analysis)
3-(2-Benzothiazolylthio)propionic acid
Formula: C₁₀H₉NO₂S₂
MW: 239,31 g/mol
Número MDL: MFCD00022878
Núm. CAS: 4767-00-4

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 147.0 to 150.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR