6-(Bromomethyl)-2-pyridinemethanol ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-(Bromomethyl)-6-(hydroxymethyl)pyridine, (6-(Bromomethyl)pyridin-2-yl)methanol
B1963-1G
B1963-5G
TCIAB1963-1GEA
174
EUR
TCIAB1963-1G
TCIAB1963-5G
6-(Bromomethyl)-2-pyridinemethanol ≥98.0% (by GC, titration analysis)
6-(Bromomethyl)-2-pyridinemethanol
Formula:
C₇H₈BrNO MW: 202,05 g/mol |
Número MDL:
MFCD01631297 Núm. CAS: 40054-01-1 |
Resultados test especificaciones
Appearance | White to Light yellow to Light red powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...