9-(1H-Benzotriazol-1-ylmethyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
B1967-1G
B1967-5G
TCIAB1967-1GEA
61
EUR
TCIAB1967-1G
TCIAB1967-5G
9-(1H-Benzotriazol-1-ylmethyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
9-(1H-Benzotriazol-1-ylmethyl)-9H-carbazole
Formula:
C₁₉H₁₄N₄ MW: 298,35 g/mol |
Número MDL:
MFCD00216631 Núm. CAS: 124337-34-4 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 186.0 to 193.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...