Bis(3-ethyl-5-methyl-4-maleimidophenyl)methane ≥98.0% (by total nitrogen basis)

Proveedor: TCI

Sinónimos: Methylenebis(3-ethyl-5-methyl-4-maleimidobenzene)

B1971-25G B1971-500G B1971-100G
TCIAB1971-25GEA 45 EUR
TCIAB1971-25G TCIAB1971-500G TCIAB1971-100G
Bis(3-ethyl-5-methyl-4-maleimidophenyl)methane ≥98.0% (by total nitrogen basis)
Bis(3-ethyl-5-methyl-4-maleimidophenyl)methane
Formula: C₂₇H₂₆N₂O₄
MW: 442,52 g/mol
Número MDL: MFCD00467754
Núm. CAS: 105391-33-1
EINECS: 424-600-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=112be2b90a2e95a16e1e711aff6669562e74bd51&vid=9f0063961a6e10766d4c8fa060fc433b1b2439dc&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 163.0 to 166.0 deg-C
Solubility in Acetonitrile almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR