Boc-L-tyrosine methyl ester ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
Boc-Tyr-OMe, N-Boc-L-tyrosine Methyl Ester
B2005-25G
B2005-5G
TCIAB2005-25GEA
134
EUR
TCIAB2005-25G
TCIAB2005-5G
Boc-L-tyrosine methyl ester ≥98.0% (by HPLC, total nitrogen)
Boc-L-tyrosine methyl ester
Formula:
C₁₅H₂₁NO₅ MW: 295,3 g/mol |
Número MDL:
MFCD00191181 Núm. CAS: 4326-36-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 104.0 to 107.0 deg-C |
Specific rotation [a]20/D | +48.0 to +52.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...