N-(tert-Butoxycarbonyl)-L-serinemethylester ≥95.0% (by HPLC, total nitrogen)

Proveedor: TCI

Sinónimos: Boc-Ser-OMe, N-Boc-L-serine Methyl Ester, BOC-L-Serine methyl ester

B2073-25G B2073-5G
TCIAB2073-25GEA 66 EUR
TCIAB2073-25G TCIAB2073-5G
N-(tert-Butoxycarbonyl)-L-serinemethylester ≥95.0% (by HPLC, total nitrogen)
N-(tert-Butoxycarbonyl)-L-serine methyl ester
Formula: C₉H₁₇NO₅
MW: 219.24 g/mol
Número MDL: MFCD00191869
Núm. CAS: 2766-43-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=819e5d9c0d3ec7fac523ef9fe36105052bed2f8c&vid=819e5d9c0d3ec7fac523ef9fe36105052bed2f8c&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Light yellow to Light orange clear liquid to slightly cloudy liquid
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Specific rotation [a]20/D -15.0 to -21.0 deg (C=5, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR