(S)-(-)-3-Benzyloxy-1,2-propanediol ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
(S)-(-)-Glycerol alpha-Benzyl Ether
B2142-1G
B2142-100MG
TCIAB2142-1GEA
195
EUR
TCIAB2142-1G
TCIAB2142-100MG
(S)-(-)-3-Benzyloxy-1,2-propanediol ≥98.0% (por GC)
(S)-(-)-3-Benzyloxy-1,2-propanediol
Formula:
C₁₀H₁₄O₃ MW: 182,22 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00077914 Núm. CAS: 17325-85-8 |
Resultados test especificaciones
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (LC) | min. 98.0 ee% |
Specific rotation [a]20/D | -4.0 to -7.0 deg (C=20, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...