Bis(methyldiphenylphosphine)palladium(II) dichloride cis- and trans-mixture ≥96.0% (by titrimetric analysis)

Proveedor: TCI
Atención

B2161-1G B2161-5G
TCIAB2161-1GEA 128 EUR
TCIAB2161-1G TCIAB2161-5G
Bis(methyldiphenylphosphine)palladium(II) dichloride cis- and trans-mixture ≥96.0% (by titrimetric analysis)
Bis(methyldiphenylphosphine)palladium(II) dichloride
Formula: C₂₆H₂₆Cl₂P₂Pd
Número MDL: MFCD00216594
Núm. CAS: 52611-08-2

Order Now

Resultados test especificaciones

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Chelometric Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR