(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone ≥98.0% (por GC)

Proveedor: TCI

B2165-1G B2165-25G B2165-5G
TCIAB2165-1GEA 30 EUR
TCIAB2165-1G TCIAB2165-25G TCIAB2165-5G
(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone ≥98.0% (por GC)
(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone
Formula: C₁₃H₁₅NO₃
MW: 233,2631 g/mol
Número MDL: MFCD00269687
Núm. CAS: 131685-53-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Melting point 43.0 to 47.0 deg-C
Specific rotation [a]20/D -95.0 to -110.0 deg (C=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR