(S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone ≥96.0% (por GC)
Proveedor: TCI
Sinónimos:
(4S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone, (S)-4-Benzyl-3-propionyloxazolidin-2-one
B2166-1G
B2166-5G
TCIAB2166-1GEA
35
EUR
TCIAB2166-1G
TCIAB2166-5G
(S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone ≥96.0% (por GC)
(S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone
Formula:
C₁₃H₁₅NO₃ MW: 233,27 g/mol |
Número MDL:
MFCD00269688 Núm. CAS: 101711-78-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Optical purity (LC) | min. 98.0 ee% |
Melting point | 43.0 to 47.0 deg-C |
Specific rotation [a]20/D | +98.0 to +108.0 deg (C=1, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...