Boc-D-Alanine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: N-Boc-D-alanine, Boc-D-Ala-OH

B2175-25G B2175-5G
TCIAB2175-25GEA 77 EUR
TCIAB2175-25G TCIAB2175-5G
Boc-D-Alanine ≥98.0% (by HPLC, titration analysis)
Boc-D-Alanine
Formula: C₈H₁₅NO₄
MW: 189,2 g/mol
Número MDL: MFCD00063123
Núm. CAS: 7764-95-6

Order Now

Resultados test especificaciones

Appearance White powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 82.0 to 85.0 deg-C
Specific rotation [a]20/D +25.0 to +27.0 deg (C=2, AcOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR