1,1'-[Biphenyl-4,4'-diylbis(methylene)]bis(4,4'-bipyridinium)dibromide ≥96.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
B2279-1G
B2279-100MG
TCIAB2279-1GEA
327
EUR
TCIAB2279-1G
TCIAB2279-100MG
1,1'-[Biphenyl-4,4'-diylbis(methylene)]bis(4,4'-bipyridinium)dibromide ≥96.0% (by HPLC, titration analysis)
1,1'-[Biphenyl-4,4'-diylbis(methylene)]bis(4,4'-bipyridinium)dibromide
Formula:
C₃₄H₂₈Br₂N₄ MW: 652,43 g/mol |
Número MDL:
MFCD06797054 Núm. CAS: 2232875-70-4 |
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...